Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Promega Immobilized Trypsin

Catalog No. PRV9012 Shop All Promega Corporation Products
Change view
Click to view available options
Kit Contents:
2 mL Immobilized Trypsin, Spin column with caps (10 Each)
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Kit Contents
PRV9012 2 mL Immobilized Trypsin, Spin column with caps (10 Each)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PRV9012 Supplier Promega Supplier No. V9012

Immobilized Trypsin provides a fast, convenient method for digesting a range of concentrations of purified protein or complex protein mixtures. Digested peptides are separated from the trypsin as they flow through the spin column into the collection tube.

  • Fast, Convenient Digestion of Purified Protein or Protein Mixtures
  • Digest a range of protein concentrations
  • Digestions completed in as little as 30 minutes

Specifications

For Use With (Application) For effective removal of Trypsin, Chymotrypsin and Elastase from Protein digests
Kit Contents 2 mL Immobilized Trypsin, Spin column with caps (10 Each)
Sufficient For 10 Reactions
Content And Storage 2°C to 10°C
Quantity 2 mL
Product Type Trypsin
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.