Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Promega ImProm-II™ Reverse Transcription

Catalog No. PRA3801 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Product Type:
ImProm-II Reverse Transcriptase
ImProm-II Reverse Transcription System
No. of Reactions:
10
100
500
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type No. of Reactions
PRA3801 ImProm-II Reverse Transcriptase 10
PRA3800 ImProm-II Reverse Transcription System 100
PR-A3802 ImProm-II Reverse Transcriptase 100
PR-A3803 ImProm-II Reverse Transcriptase 500
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. PRA3801 Supplier Promega Supplier No. A3801
Add to Cart
Add to Cart
This item is not returnable. View return policy

Formulated for stand-alone applications or for easy transition to gene-specific target amplification.

  • Reverse transcribes RNA templates from total RNA, poly(A)+ mRNA, or synthetic transcript RNA
  • Enables robust, full-length cDNA synthesis for the analysis of rare or long messages (up to 8.9kb)
  • Uses volumes of 1–20μL from the initial RT reaction in coupled or uncoupled PCR reactions
  • May also be used for the incorporation of regular, Cy™3-modified, Cy5-modified, and amino-allyl-modified nucleotides for microarray applications
  • Refill components available separately

Specifications

Content And Storage -30°C to -10°C
For Use With (Application) Reverse transcription of RNA templates from total RNA, poly(A)+ mRNA, or synthetic transcript RNA
Includes MgCl2 750μL, ImProm-II 5X reaction buffer 100μL, ImProm-II reverse transcriptase 10μL
No. of Reactions 10
pH 8.3
Product Type ImProm-II Reverse Transcriptase
Form Liquid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.