Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Promega InsR Kinase Enzyme System

Catalog No. PRV3901 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Quantity:
1/Ea.
10 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PRV3901 10 μg
PRV9411 1/Ea.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. PRV3901 Supplier Promega Supplier No. V3901
Add to Cart
Add to Cart

Includes

InsR Kinase Enzyme System (V3901)

  • Kinase: InsR, 10μg (Human, recombinant; amino acids 1011–end). MW: ∽70kDa
  • Substrate: Axltide (KKSRGDYMTMQIG); derived from the mouse Insulin receptor substrate 1 (amino acid 979–989)
  • Reaction Buffer, DTT, MnCl2
ADP-Glo™ Kinase Assay and EGFR Kinase Enzyme System (V9411)
  • 1 x 10μg InsR Kinase Enzyme System (V901) and 1 x 1000 ADP-Glo™ Kinase Assays (V9101)

Recombinant human InsR (amino acids 1011-end) was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag. InsR is the insulin receptor tyrosine kinase that is involved in insulin signaling.

  • Easily Screen and Profile InsR Kinase Inhibitors
  • Includes kinase, substrate and reaction buffer
  • Use with ADP-Glo™ Assay for bioluminescent detection of kinase activity

Specifications

Content And Storage <-65°C
For Use With (Application) Involved in Insulin signaling, binding of Insulin to the InsR stimulates Glucose uptake, Insulin receptor signaling helps to maintain fuel homeostasis and prevent diabetes
Quantity 10 μg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.