Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Promega ITK Kinase Enzyme System

Catalog No. PRV3191 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Quantity:
1/Ea.
10 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PRV3191 10 μg
PRV9431 1/Ea.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. PRV3191 Supplier Promega Supplier No. V3191
Add to Cart
Add to Cart

Includes

ITK Kinase Enzyme System (V3191)

  • Kinase: ITK, 10μg (Human, recombinant; amino acids 352–end), M.W.: ∽53kDa
  • Substrate: Poly (4:1 Glu, Tyr) Peptide
  • Others: Reaction Buffer, DTT, MnCl2
ADP-Glo™ Kinase Assay and ITK Kinase Enzyme System (V9261)
  • 1 x 10μg ITK Kinase Enzyme System (V3191) and 1 x 1000 ADP-Glo™ Kinase Assays (V9261)

Recombinant human ITK (amino acids 352-end) was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag. ITK is a member of the TEC family of nonreceptor tyrosine kinases.

  • Easily Screen and Profile ITK Kinase Inhibitors
  • Includes kinase, substrate and reaction buffer
  • Use with ADP-Glo™ Assay for bioluminescent detection of kinase activity

Specifications

Molecular Weight (g/mol) 53
Content And Storage <-65°C
Quantity 10 μg
Form Fluid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.