Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Promega JAK3 Kinase Enzyme System

Catalog No. PRV3701 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Quantity:
1/Ea.
10 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PRV3701 10 μg
PRV9441 1/Ea.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. PRV3701 Supplier Promega Supplier No. V3701
Add to Cart
Add to Cart

Includes

JAK3 Kinase Enzyme System (V3701)

  • 10μg of JAK3 Kinase (Human, recombinant; amino acids 781–end)
  • Poly (4:1 Glu4,Tyr1) peptide substrate
  • Reaction buffer
  • DTT
ADP-Glo™ Kinase Assay and JAK3 Kinase Enzyme System (V9441)
  • 1 x 10μg JAK3 Kinase Enzyme System (V3831) and 1 x 1000 ADP-Glo™ Kinase Assays (V9101)

Recombinant human JAK3 (amino acids 781-end) was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag. JAK3 is a member of the JAK family of tyrosine kinases involved in cytokine receptor-mediated intracellular signal transduction.

  • Easily Screen and Profile JAK3 Kinase Inhibitors
  • Includes kinase, substrate and reaction buffer
  • Use with ADP-Glo™ Assay for bioluminescent detection of kinase activity

Specifications

Content And Storage <-65°C
For Use With (Application) Involved in Cytokine receptor-mediated intracellular signal transduction, differentiation of hematopoietic cells progression of B-cell development in the Bone Marrow
Quantity 10 μg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.