Learn More
Description
- Use with 5', 3' or blunt-ended DNA inserts
- Ligation of cohesive ends in 5 minutes, blunt ends in 15 minutes at room temperature
- No requirement to purify ligated DNA prior to heat-shock transformation in E. coli ligations conducted at room temperature
- No additional buffer modifications required prior to use
- Ligations performed using the LigaFast™ system are comparable to standard overnight ligations
- Blue/White Cloning Qualified
Specifications
Specifications
| Content And Storage | -30°C to -10°C |
| Formulation | Tris HCl (pH 7) 10mM, KCl 50mM, DTT 1mM, EDTA 0.1mM, Glycerol 50% |
| For Use With (Application) | 5′, 3′ or blunt ended DNA inserts |
| pH | 7.8 |
| Product Type | DNA Ligation System |
| Quantity | 30 Reactions |
| Form | Fluid |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.