Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Promega M-MLV Reverse Transcriptase, RNase H Minus

Catalog No. PRM5301 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Quantity:
10,000 U
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PRM5301 10,000 U
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PRM5301 Supplier Promega Supplier No. M5301
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Deletion mutation of M-mLV RT resulting in no RNase H activity. Suitable for rare or long transcripts.

  • Helpful for Preparing Long cDNAs and Libraries with a High Percentage of Full-Length cDNA
  • Supplied at a concentration of 100-200u/μL

Specifications

Concentration 100 to 200 U/μL
Color Colorless
Components 5X Reaction Buffer: 250mM Tris HCl (pH 8.3 at 25°C), 375mM KCl, 15mM MgCl2, 50mM DTT
Purity >90%
pH 7.5
For Use With (Application) First-strand cDNA synthesis, primer extension
Storage Buffer 20mM Tris HCl (pH 7.5 at 25°C), 0.2M NaCl, 0.1mM EDTA, 1mM DTT, 0.01% Nonidet™ P-40, 50% Glycerol
Content And Storage -30°C to -10°C
Source Recombinant E. coli strain
Quantity 10,000 U
Product Type M-MLV Reverse Transcriptase, RNase (H-)
Inhibitors Recombinant RNasin™ Ribonuclease
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.