Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Promega M-MLV Reverse Transcriptase, RNase H Minus, Point Mutant

Catalog No. PRM3683 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Description:
M-MLV Reverse Transcriptase
Quantity:
10000U
2500U
50000U
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Description Quantity
PRM3683 M-MLV Reverse Transcriptase 50000U
PRM3682 M-MLV Reverse Transcriptase 10000U
PRM3681 M-MLV Reverse Transcriptase 2500U
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. PRM3683 Supplier Promega Supplier No. M3683
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Point mutation of M-MLV RT resulting in no RNase H activity. Suitable for rare or long transcripts.

  • High Yields of cDNA from Long RNA Templates (>5kb)

Specifications

Content And Storage −30 to −10°C
Description M-MLV Reverse Transcriptase
Form Liquid
Quantity 50000U
For Use With (Application) cDNA Synthesis
Includes 5 x 1mL M-MLV RT 5X Buffer
Source Recombinant E. coli Strain
Storage Buffer Tris-HCl (pH 7.5 at 25°C) 20mM, NaCl 200mM, EDTA 0.1mM, DTT 1mM, 0.01% Nonidet™ P40™ and Glycerol 50%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.