Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Promega Maxwell™ 16 Purification Kits

Catalog No. PRAS1120
Click to view available options
Product Type:
Blood DNA Kit
Cell DNA Kit
LEV Blood DNA Kit
Mouse Tail Tissue DNA Kit
Tissue DNA Kit
Viral Total Nucleic Acid Kit

Includes

All kits include 48 Maxwell kit-specific cartridges, 50 purification plungers, 50 elution tubes, and 20mL elution buffer. FFPE Tissue LEV DNA Purification Kit also includes 32mL lysis buffer, 2 x 10mg proteinase K, and 15mL incubation buffer. Viral Total Nucleic Acid Kit also includes 20mL lysis buffer, 2 x 1mL proteinase K solution, and 20mL nuclease-free water. LEV Blood DNA Kit also includes 20mL lysis buffer and 2 x 1mL proteinase K solution.

Requires

Maxwell 16 DNA Purification Instrument (Mfr. No. AS1120 not for use with Maxwell 16 MDx); LEV Kits also require the LEV Hardware Kit (Mfr. No. AS1250)

Used with Maxwell 16 Instruments to provide an easy method for efficient, automated purification of genomic DNA

  • Automated DNA Extraction from Mouse Tails, Ear Punches and Other Tissue
  • Yields up to 20μg of DNA from 1.2cm of tissue
  • Eliminates grinding or Proteinase K treatment

  • U.S. Patents 6,027,945 and 6,368,800; Australian Pat. No. 732756.
  • Mfr. Nos. AS1010, AS1020, AS1030, and AS1140: This product is licensed for use under U.S. Patent 6,174,704.
  • Mfr. Nos. AS1130, AS1140: U.S. Patent 6,673,631.
  • Mfr. Nos. AS1130, AS1140: Patent pending.

Specifications

Content And Storage 15°C to 30°C
For Use With (Equipment) Maxwell 16 Instrument
For Use With (Application) Automated genomic DNA purification from mouse tail tissue
Includes 48 Maxwell™ 16 Mouse tail DNA cartridges, 50 purification plungers, 50 Elution tubes, 20mL Elution Buffer
No. of Reactions 48 Preps
Product Type Mouse Tail Tissue DNA Kit
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.