Learn More
Promega MET Kinase Enzyme System
Catalog No. PRV3361 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Quantity:
1/Ea.
10 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PRV3361 10 μg
PRV9571 1/Ea.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. PRV3361 Supplier Promega Supplier No. V3361
Only null left
Add to Cart
Add to Cart
Includes
MET Kinase Enzyme System (V3361)
  • Kinase: MET, 10μg (Human, recombinant; amino acids 352–end), M.W.: ∽81kDa
  • Substrate: Poly (4:1 Glu, Tyr) Peptide
  • Others: Reaction Buffer, DTT
ADP-Glo™ Kinase Assay and MET Kinase Enzyme System (V9571)
  • 1 x 10μg MET Kinase Enzyme System (V3361) and 1 x 1000 ADP-Glo™ Kinase Assays (V9571)
Recombinant human MET (amino acids 956-end) was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag. MET is a proto-oncogene that encodes a transmembrane growth factor receptor.
  • Easily Screen and Profile MET Kinase Inhibitors
  • Includes kinase, substrate and reaction buffer
  • Use with ADP-Glo™ Assay for bioluminescent detection of kinase activity
Specifications
Content And Storage <-65°C
Quantity 10 μg