Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Promega MgCl2 Solution, 25 mM

Catalog No. PRA3511 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Quantity:
1.5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PR-A3511 1.5 mL
PR-A3513 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. PR-A3511 Supplier Promega Supplier No. A3511
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Magnesium Chloride Solution, supplied at 25mM, is used with GoTaq™ Flexi Reaction Buffers for PCR optimization.

  • High Quality Reagent for Optimal PCR
  • MgCl2 solution supplied at 25mM
  • Use with GoTaq(R) Flexi Reaction Buffers for PCR optimization

Specifications

Concentration 25 mM
Content And Storage 2 x 750 μL MgCl2. Store at –30° to –10°C.
Quantity 1.5 mL
For Use With (Application) RT-PCR
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.