Learn More
Description
- Characterize MSI Status of Your Research Samples
- Greater accuracy than immunohistochemistry (IHC) for identifying MSI-high samples
- Fluorescent Multiplex PCR-based method using DNA extracted from precious samples
- Easier to use, less expensive and faster turnaround than NGS
Specifications
Specifications
| Content And Storage | DNA high molecular weight (1 x 3μg), Internal lane standard 600 (1 x 150μL), Gold Star 10X Buffer (1 x 300μL), MSI 10X primer pair mix (1 x 100μL), Nuclease free Water (1 x 1250μL). Store at −30 to −10°C |
| Quantity | 50 Pr. |
| For Use With (Application) | Microsatellite Instability Analysis |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.