Learn More
Promega Monster Green™ Fluorescent Protein phMGFP Vector
Catalog No. pre6421
Click to view available options
Quantity:
20 μg
The Monster Green Fluorescent Protein encoded by the hMGFP gene is an ideal fluorescent reporter, providing high-level fluorescence and reducing cytotoxicity.
  • Improved Synthetic GFP Reporter with Greater Fluorescent Intensity and Reduced Cytotoxicity
  • Visualize low-level expression in situ using fluorescence microscopy, imagers or FACS(R)
  • Create fusion proteins for imaging and localization studies using standard FITC detection
  • Transfect at high efficiencies for precloning confirmation studies
  • Australian Pat. No. 2003297293 and other patents pending.
  • The gene encoding Monster Green Fluorescent Protein is exclusively licensed to Promega Corporation under U.S. Pat. Nos. 7,291,711 and 7,413,874 and other patents pending.
  • Certain applications of this product may require licenses from others.
  • For research use only. Researchers may use this product obtained from Promega for their own research and they may transfer derivatives of this product to their research colleagues who agree in writing to be bound by the terms and conditions of this label license. No transfer of unmodified product is authorized. Research performed on behalf of another party is permitted. No other use or transfer of this product or its derivatives is authorized without the express, written consent of Promega. Such prohibition includes, without limitation, any and all nonresearch uses of this product in any procedure, application, product manufacture, or product transfer for which monetary or other consideration is received from another party. With respect to such commercial use, or any therapeutic or diagnostic uses, please contact Promega Corporation for supply and licensing information.
Specifications
Promoter CMV
Content And Storage -30°C to -10°C
Quantity 20 μg
Vector phMGFP vector
For Use With (Application) Provides high-purity for high transfection efficiencies, reduces cytotoxicity, offering flexibility when working with transient and stable expression assays
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.