Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Promega NAD(P)H-Glo™ Detection System

Catalog No. PRG9061 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Quantity:
10 mL
50 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PRG9061 10 mL
PRG9062 50 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. PRG9061 Supplier Promega Supplier No. G9061
Add to Cart
Add to Cart

The NAD(P)H-Glo™ Detection System is a bioluminescent assay that measures the concentration of the reduced forms of nicotinamide adenine dinucleotides, NADH and NADPH, and does not discriminate between them.

  • Monitor the Concentration of Reduced Forms of NADH and NADPH
  • Highly sensitive and rapid bioluminescent assay
  • Easily adapted for high-throughput format

Specifications

Quantity 10 mL
Content And Storage -20°C
Detection Method Bioluminescence
Kit Contents 1 x 55μL Reductase, 1 x 55μL reductase substrate, 1 x 1 Luciferin detection Reagent, 1 x 10mL Reconstitution Buffer
pH 7.5
Form Fluid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.