Learn More
Description
- Monitor the Concentration of Reduced Forms of NADH and NADPH
- Highly sensitive and rapid bioluminescent assay
- Easily adapted for high-throughput format
Specifications
Specifications
| Quantity | 50 mL |
| Content And Storage | 1 x 275μL Reductase, 1 x 275μL reductase substrate, 1 x 1 Luciferin detection Reagent, 1 x 50mL Reconstitution Buffer -20°C |
| Detection Method | Bioluminescence |
| pH | 7.5 |
| Form | Fluid |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.