Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Promega NanoLuc™ pNL3 Vectors

Catalog No. PRN1031 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Product Type:
pNL3.1(Nluc/minP)
pNL3.2.NF-κB-RE(NlucP/NF-κB-RE/Hygro)
pNL3.3(secNluc/minP)
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
PRN1031 pNL3.1(Nluc/minP)
PRN1051 pNL3.3(secNluc/minP)
PRN1111 pNL3.2.NF-κB-RE(NlucP/NF-κB-RE/Hygro)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. PRN1031 Supplier Promega Supplier No. N1031
Add to Cart
Add to Cart

Configured with NanoLuc Luciferase for use in reporter gene assays of transcriptional control—ideal for cloning of binding site or response element not in need of basic promoters

  • Clone Response Elements of Interest to Drive Expression of NanoLuc(R) Luciferase
  • Nluc genes are codon optimized for mammalian cells
  • Many potential regulatory elements or other undesirable features have been removed (such as common restriction enzyme sites)
  • Vectors have a pGL4-based backbone for easy sequence transfer from existing plasmids

Specifications

Product Type pNL3.1(Nluc/minP)
pH 8
Content And Storage -30°C to -10°C
Quantity 20 μg
Form Liquid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.