Learn More
Description
- Enhances Protein Expression in Mammalian Cells When Cotransfected
- Increases translation initiation in cells transiently transfected with protein expression constructs
- Can be used in a variety of cell lines
Specifications
Specifications
| Storage Buffer | TE Buffer (10mMTris, pH 7.5, 0.1mM EDTA) |
| Content And Storage | -30°C to -10°C |
| Quantity | 20 μg |
| Vector | pAdVAntage vector |
| For Use With (Application) | Enhances transient Protein expression in a variety of cell types by increasing translation initiation, shows tenfold increase in Luciferase expression in 293 and HeLa cell lines over transfections performed with the construct DNA alone |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.