Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Promega PDGFRα Kinase Enzyme System

Catalog No. PRV3721 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Quantity:
1/Ea.
10 μg
Product Type:
ADP-Glo™ Kinase Assay, PDGFRα Kinase Enzyme System
PDGFRα Kinase Enzyme System
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Product Type
PRV3721 10 μg PDGFRα Kinase Enzyme System
PRV8011 1/Ea. ADP-Glo™ Kinase Assay, PDGFRα Kinase Enzyme System
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. PRV3721 Supplier Promega Supplier No. V3721
Add to Cart
Add to Cart

Includes

  • Kinase: PDGFRα, 10μg (Human, recombinant; amino acids 550–end), M.W.: ∽95kDa
  • Substrate: Poly (4:1 Glu, Tyr) Peptide
  • Others: Reaction Buffer, DTT

Recombinant human PDGFR alpha (amino acids 550-end) was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag. PDGFR alpha is a member of the PDGFR family of membrane receptors with intrinsic tyrosine kinase activity.

  • Easily Screen and Profile PDGFRα Kinase Inhibitors
  • Includes kinase, substrate and reaction buffer
  • Use with ADP-Glo™ Assay for bioluminescent detection of kinase activity

Specifications

Product Type PDGFRα Kinase Enzyme System
Molecular Weight (g/mol) 95
Content And Storage <-65°C
Quantity 10 μg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.