Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Promega PDK1 Kinase Enzyme System

Catalog No. PRV9681 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Quantity:
1/Ea.
10 μg
Content And Storage:
<−65°C
COMPONENTS AT DIFFERENT TEMPS
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Content And Storage
PRV9681 1/Ea. COMPONENTS AT DIFFERENT TEMPS
PRV2761 10 μg <−65°C
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. PRV9681 Supplier Promega Supplier No. V9681
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Recombinant full-length human PDK1 was expressed by baculovirus in Sf9 insect cells using an N-terminal His tag. PDK1 (3-phosphoinositide-dependent protein kinase) is activated by the presence of PtdIns(3,4,5)P3 or PtdIns(3,4)P2.

  • Easily Screen and Profile PDK1 Kinase Inhibitors
  • Includes kinase, substrate and reaction buffer
  • Use with ADP-Glo™ Assay for bioluminescent detection of kinase activity

Specifications

Includes 10μg Recombinant PDK1 Kinase, PDKtide (KTFCGTPEYLAPEVRREPRILSEEEQEMFRDFDYIADWC) Substrate, Reaction Buffer, DTT and MnCl2
Molecular Weight (g/mol) 67
Content And Storage COMPONENTS AT DIFFERENT TEMPS
Product Type PDK1 Kinase Enzyme system
Quantity 1/Ea.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.