Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Promega pFC30A His6HaloTag™ T7 Flexi Vector for E. coli and Cell-Free Protein Expression

Catalog No. PRG8321
Click to view available options
Quantity:
20 μg

These vectors are used for inducible expression of polyhistidine-HaloTag™ fusion proteins in E. coli and cell-free systems using the T7 RNA polymerase promoter. Use the Flexi Cloning System vectors for C- or N-terminal fusions.

  • HaloTag(R)-Polyhistidine Dual Tag Bacterial Expression Vectors
  • Express proteins in E. coli T7 cell-free expression systems
  • Couple the protein solubility and labeling benefits of HaloTag(R) technology with the reusability and the throughput of Ni-affinity technology
  • Select N- or C-terminal tags and either Ampicillin or Kanamycin resistance

Specifications

Kit Contents C-terminal His6-HaloTag™ dual tag
Type pFC30A His-6HaloTag T7 Flexi Vector
Sufficient For E. coli and T7 cell-free Protein expression systems
Quantity 20 μg
Content And Storage -30°C to -10°C
For Use With (Application) Bacterial Protein expression and purification, cell-free Protein expression and purification
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.