Learn More
Description
- Firefly Luciferase Reporter Vectors with Cellular Stress Response Elements
- Offer a range of response elements that are involved in stress signalling pathways
- Each vector included an optimized firefly luciferase gene that codes a reporter with a one-hour half-life
- Hygromycin resistance selectable marker means choosing transient transfection or generating a stable cell line
Specifications
Specifications
| Marker Gene | Hygro |
| Promoter | SV40 |
| Protein Degradation Sequence | hPEST |
| Reporter Gene | luc2P |
| Storage Buffer | 10mM Tris HCl (pH 7.4), 1mM EDTA |
| Content And Storage | -30°C to -10°C |
| Concentration | 1 μg/μL |
| Primer | RV Primer 3 and RV Primer 4 |
| Quantity | 20 μg |
| Vector | pGL4.39[luc2P/ATF6 RE/Hygro] |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.