Learn More
Description
- Cytokine Response Elements in Firefly Luciferase Reporter Vectors
- Offer a range of response elements that are involved in cytokine signalling pathways
- Each vector included an optimized firefly luciferase gene that codes a reporter protein with a one-hour half-life
- Hygromycin resistance selectable marker means choosing transient transfection or generating a stable cell line
Specifications
Specifications
| Marker Gene | Hygro |
| Promoter | SV40 |
| Protein Degradation Sequence | hPEST |
| Reporter Gene | luc2P |
| Storage Buffer | 10mM Tris HCl (pH 7.4), 1mM EDTA |
| Content And Storage | -30°C to -10°C |
| Concentration | 1 μg/μL |
| Primer | RV Primer 3 and RV Primer 4 |
| Quantity | 20 μg |
| Vector | pGL4.48[luc2P/SBE/Hygro] |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.