Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Promega pGL4 Minimal Promoter Driven Luciferase Vectors

Catalog No. PRE3751 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Vector:
pGL4.38[luc2P/p53 RE/Hygro]
pGL4.39[luc2P/ATF6 RE/Hygro]
pGL4.40[luc2P/MRE/Hygro]
pGL4.41[luc2P/HSE/Hygro]
pGL4.45[luc2P/ISRE/Hygro]
pGL4.47[luc2P/SIE/Hygro]
pGL4.48[luc2P/SBE/Hygro]
pGL4.49[luc2P/TCF-LEFRE/Hygro]
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Vector
PRE3751 pGL4.41[luc2P/HSE/Hygro]
PRE3651 pGL4.38[luc2P/p53 RE/Hygro]
PRE3671 pGL4.48[luc2P/SBE/Hygro]
PRE4041 pGL4.47[luc2P/SIE/Hygro]
PRE4131 pGL4.40[luc2P/MRE/Hygro]
PRE4141 pGL4.45[luc2P/ISRE/Hygro]
PRE4611 pGL4.49[luc2P/TCF-LEFRE/Hygro]
PRE3661 pGL4.39[luc2P/ATF6 RE/Hygro]
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Catalog No. PRE3751 Supplier Promega Supplier No. E3751
Add to Cart
Add to Cart

Firefly luciferase reporter vectors with response elements.

  • Firefly Luciferase Reporter Vectors with Cellular Stress Response Elements
  • Offer a range of response elements that are involved in stress signalling pathways
  • Each vector included an optimized firefly luciferase gene that codes a reporter with a one-hour half-life
  • Hygromycin resistance selectable marker means choosing transient transfection or generating a stable cell line

Specifications

Marker Gene Hygro
Promoter SV40
Protein Degradation Sequence hPEST
Reporter Gene luc2P
Storage Buffer 10mM Tris HCl (pH 7.4), 1mM EDTA
Content And Storage -30°C to -10°C
Concentration 1 μg/μL
Primer RV Primer 3 and RV Primer 4
Quantity 20 μg
Vector pGL4.41[luc2P/HSE/Hygro]
For Use With (Application) Transcription regulation, virus-cell interactions, compound screening, post-translational modifications, GPCR signaling, cell signaling, promoter analysis, transient transfection experiments
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.