Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Promega pGL4 Minimal Promoter Driven Luciferase Vectors

Catalog No. pre3651
Click to view available options
Vector:
pGL4.38[luc2P/p53 RE/Hygro]
pGL4.39[luc2P/ATF6 RE/Hygro]
pGL4.40[luc2P/MRE/Hygro]
pGL4.41[luc2P/HSE/Hygro]
pGL4.45[luc2P/ISRE/Hygro]
pGL4.47[luc2P/SIE/Hygro]
pGL4.48[luc2P/SBE/Hygro]
pGL4.49[luc2P/TCF-LEFRE/Hygro]

Firefly luciferase reporter vectors with response elements.

  • Firefly Luciferase Reporter Vectors with Cellular Stress Response Elements
  • Offer a range of response elements that are involved in stress signalling pathways
  • Each vector included an optimized firefly luciferase gene that codes a reporter with a one-hour half-life
  • Hygromycin resistance selectable marker means choosing transient transfection or generating a stable cell line

Specifications

Marker Gene Hygro
Promoter SV40
Protein Degradation Sequence hPEST
Reporter Gene luc2P
Storage Buffer 10mM Tris HCl (pH 7.4), 1mM EDTA
Content And Storage -30°C to -10°C
Concentration 1 μg/μL
Primer RV Primer 3 and RV Primer 4
Quantity 20 μg
Vector pGL4.38[luc2P/p53 RE/Hygro]
For Use With (Application) Transcription regulation, virus-cell interactions, compound screening, post-translational modifications, GPCR signaling, cell signaling, promoter analysis, transient transfection experiments
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.