Learn More
Promega PKCζ Kinase Enzyme System
Catalog No. PRV2781 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Quantity:
1/Ea.
10 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PRV2781 10 μg
PRV9731 1/Ea.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. PRV2781 Supplier Promega Supplier No. V2781
Add to Cart
Add to Cart
Includes
PKCζ Kinase Assay:
  • Kinase: Recombinant Human PKCζ, 10μg (full length), MW: ≅93kDa
  • Substrate: CREBtide (KRREILSRRPSYR) derived from human CREB1 isoform A (amino acid 109 to 121)
  • Other: Reaction Buffer, DTT
ADP-Glo Kinase Assay: Reagent, buffer, substrate, Ultra Pure ATP (10mM), ADP (10mM)
Recombinant full-length human PKC zeta was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag. PKC zeta is an atypical isoform of the PKC family.
  • Easily Screen and Profile PKCζ Kinase Inhibitors
  • Includes kinase, substrate and reaction buffer
  • Use with ADP-Glo™ Assay for bioluminescent detection of kinase activity
Specifications
Content And Storage <-65°C
Quantity 10 μg