Learn More
Description
• Measures protein kinase A activity in a multiwell plate format using fluorescence intensity readout
• Homogeneous assay is performed in a single well
• Highly predictive Z values are >0.8 in 96– and 384–well plates
• No radioactive waste or disposal issues
- Patent pending.
- This product is covered by U.S. Patents 4,557,862 and 4,640,893 and is sold for research use only. All other used, including, but not limited to, use as a clinical diagnostic or therapeutic, require a separate license. Please contact Promega Corporation for details.
Specifications
Specifications
| For Use With (Application) | Measures Protein Kinase A activity in a multiwell plate format using Fluorescence intensity readout |
| Content And Storage | -20°C |
| Product Type | PKA assay system |
| Quantity | 4 Plates |
| Format | 96 or 384-well Plate |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.