Learn More
Promega ProFluor™ SerThr Phosphatase Assay
Catalog No. PRV1261 Shop All Promega Corporation Products
Change view
Click to view available options
Quantity:
4 Plate
8 Plate
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PRV1261 8 Plate
PRV1260 4 Plate
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. PRV1261 Supplier Promega Supplier No. V1261
The ProFluor Ser/Thr PPase Assay measures purified serine/threonine protein phosphatase activity in a multiwell plate format and is ideal for high-throughput applications.
  • Fluorescent, “Add-Mix-Read” Ser/Thr Phosphatase Assay
  • Works with protein phosphatase 1 (PP1), PP2A, PP2B and PP2C
  • Ideal for high-throughput phosphatase assays in 96- or 384-well plate formats
  • Minimal test compound interference
Patent pending. This product is covered by U.S. Patents 4,557,862 and 4,640,893 and is sold for research use only. All other uses, including, but not limited to, use as a clinical diagnostic or therapeutic, require a separate license. Please contact Prom
Specifications
Description ProFluor Ser/Thr Assay System
Content And Storage −30 to −10°C
Quantity 8 Plate