Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Promega ProFluor™ Src-Family Kinase Assay Systems

Catalog No. PRV1270 Shop All Promega Corporation Products
Change view
Click to view available options
Quantity:
4 Plates
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PRV1270 4 Plates
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PRV1270 Supplier Promega Supplier No. V1270

ProFluor Src-Family Kinase Assay measures the activity of purified Src-family tyrosine kinases in multiwell plate format and is ideal for high-throughput applications.

  • Fluorescence-Based Detection of Src-Family Tyrosine Kinase Activity
  • Robust Z´ values greater than 0.7 in either 96- or 384-well plate formats
  • Add-mix-read format reduces the number of plate-handling steps
  • Non-Radioactive: No radioactive waste disposal and safety issues

Specifications

For Use With (Application) Measures the activity of purified Src-family Tyrosine Kinases in 96 or 384-well plates, ideal for high throughput applications
Content And Storage −30 to −10°C
Product Type Src Family Protein Kinase assay
Quantity 4 Plates
Format 96 or 384-well Plate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.