Learn More
Description
- Fluorescent, “Add-Mix-Read” Tyrosine Phosphatase Assay
- Tyrosine phosphatases tested include PTP-1B, CD45, LAR PTPase and YOP-51
- Ideal for high-throughput applications in 96- or 384-well plate formats
- Minimal test compound interference
Specifications
Specifications
| For Use With (Application) | Measures the activity of purified Tyrosine Phosphatases in 96 or 384-well plates, ideal for high throughput applications |
| Content And Storage | −30 to −10°C |
| Product Type | Tyrosine Phosphatase assay |
| Quantity | 8 Plates |
| Format | 96 or 384-well Plate |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.