Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Promega Proteasome-Glo™ Assays

Catalog No. PRG8632 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Product Type:
Caspase-like Assay
Trypsin-like Assay
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
PRG8632 Trypsin-like Assay
PRG8642 Caspase-like Assay
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. PRG8632 Supplier Promega Supplier No. G8632
Add to Cart
Add to Cart
This item is not returnable. View return policy

Includes

Buffer, substrate and luciferin detection reagent

The Proteasome-Glo™ Cell-Based Assays are homogeneous, luminescent assays that measure the chymotrypsin-like, trypsin-like and caspase-like protease activities associated with the proteasome complex in a purified enzyme preparation.

  • Measure Chymotrypsin-, Trypsin- and Caspase-Like Activities
  • Homogeneous, luminescent enzyme-based assays
  • Simple add-mix-measure protocol
  • Results in 10-30 minutes after reagent addition

Specifications

Buffer Proteasome-Glo™ cell-based buffer
Product Type Trypsin-like Assay
Quantity 50 mL
Includes Proteasome-Glo™ cell-based Buffer, Z-nLPnLD-Glo™ Substrate and Luciferin detection reagent
Content And Storage −30 to −10°C
For Use With (Application) Measures Trypsin-like Protease activity associated with the Proteasome in a purified Enzyme-based format
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.