Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Promega QuantiLum™ Recombinant Luciferase

Catalog No. PRE1702 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Quantity:
1 mg
5 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PRE1702 5 mg
PRE1701 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. PRE1702 Supplier Promega Supplier No. E1702
Add to Cart
Add to Cart

QuantiLum Recombinant Luciferase is a luciferase expressed from a cloned gene from the North American firefly (Photinus pyralis).

  • Reliable and Dependable Recombinant Luciferase
  • Available in bulk for large orders
  • Custom options available
  • Supplied at a concentration of 10-15mg/ml

The method of recombinant expression of Coleoptera luciferase is covered by U.S. Patents 5,583,024; 5,674,713; and 5,700,673.

Specifications

Quantity 5 mg
Content And Storage <-65°C
Concentration 10 to 15 mg/mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.