Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Promega Reagents for ReliaPrep™ 96 gDNA Miniprep HT System

Catalog No. PRA2641 Shop All Promega Corporation Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 mL
60 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PRA2641 60 mL
PRA2651 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. PRA2641 Supplier Promega Supplier No. A2641
Add to Cart
Add to Cart

Tris-EDTA and Tris-HCl buffer solutions that are quality tested and certified free of DNase and RNase activity.

  • Quality tested and certified free of DNase and RNase activity
  • Component of Maxwell(R) HT 96 gDNA Blood Isolation System (Cat.# A2670, A2671)

Specifications

Content And Storage 15°C to 30°C
For Use With (Equipment) ReliaPrep™ 96 genomic DNA Miniprep HT System
Quantity 60 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.