Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Promega Alcohol Wash

Catalog No. PRMD1392 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Quantity:
25 mL
120 mL
160 mL
Product Type:
Alcohol Wash, Blood
Lysis Buffer, Blood
Paramag Particles, Fixed Yield
Paramagnetic Particles
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Product Type
PRMD1392 160 mL Lysis Buffer, Blood
PRMD1441 25 mL Paramagnetic Particles
PRMD1412 120 mL Alcohol Wash, Blood
PRMD1451 25 mL Paramag Particles, Fixed Yield
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. PRMD1392 Supplier Promega Supplier No. MD1392
Add to Cart
Add to Cart

Alcohol Wash for MagneSil Blood Genomic, Max Yield System (Cat.# MD1360), and MagneSil ONE, Fixed Yield Blood Genomic System (Cat.# MD1370).

  • Lysis Buffer Used With Automated Blood DNA Purification Systems
  • Component of the MagneSil Blood Genomic, Max Yield System (Cat.# MD1360), and MagneSil ONE, Fixed Yield Blood Genomic System (Cat.# MD1370).

†Also supplied with Mfr. No. A4080.

Specifications

Content And Storage 15°C to 30°C
For Use With (Equipment) Magnesil™ Blood Genomic system, Magnesil™ one system
For Use With (Application) Automated high-throughput DNA purification from Anti-coagulated whole Blood
Product Type Lysis Buffer, Blood
Quantity 160 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.