Learn More
Description
- Reagent Used in Plasmid DNA Purification Protocols
- Cat.# A7112 is a component of the Wizard(R) Plus DNA Purification Systems
- Cat.# A7115 is a component of the PureYield™ Plasmid Midiprep System
- Cat.# A7113 and A7118 are components of the Wizard(R) SV 96 Plasmid DNA Purification System
- Cat.# A7114 is a component of the Wizard(R) MagneSil(R) Plasmid Purification System
Specifications
Specifications
| Content And Storage | 15°C to 30°C |
| For Use With (Equipment) | Wizard™ Plus DNA purification system |
| For Use With (Application) | Plasmid DNA purification by Alkaline Lysis |
| Product Type | Cell Resuspension Solution |
| Quantity | 150 mL |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.