Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Promega Restriction Enzymes - Sma I Blue/White Cloning and Genome Qualified (J)

Catalog No. PRR6121 Shop All Promega Corporation Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
1000 U
Concentration:
8 to 12 U/μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Concentration
PR-R6121 1000 U 8 to 12 U/μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PR-R6121 Supplier Promega Supplier No. R6121
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

XmaI is a neoschizomer of SmaI. Available at high concentration, containing 5,000 units of SmaI at a concentration of 40-80u/μL. (CCC/GGG GGG\CCC)

  • Blue/white cloning qualified, providing a higher level of quality control for enzymes used in cloning applications
  • Genome qualified to ensure optimal performance in genomic analysis applications
  • Available at high concentration, containing 5,000 units of SmaI at a concentration of 40-80u/μL

‡Restriction enzyme buffer included indicated in parentheses; Cloning = Blue/White Cloning Qualified; Genome = Genome Qualified.

Specifications

Concentration 8 to 12 U/μL
Components 1 x 1mL 10X Buffer J, 1 x 150μL Bovine Serum Albumin, Acetylated, 1 x 1000U SmaI, 1 x 0.25mL Multi-Core™ 10X Buffer
Incubator Temperature 37°C
pH 7.4
For Use With (Application) Provides a higher level of quality control for Enzymes used in cloning applications
Storage Buffer 10mM Tris HCl (pH 7.4), 300mM KCl, 0.1mM EDTA, 1mM DTT, 0.5mg/mL BSA, 50% Glycerol
Content And Storage -30°C to -10°C
Quantity 1000 U
Cut Site CCC.GGG GGG.CCC
Product Type SmaI
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.