Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Promega Restriction Enzymes - Stu I (B)

Catalog No. PRR6421 Shop All Promega Corporation Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
400 U
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PR-R6421 400 U
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PR-R6421 Supplier Promega Supplier No. R6421

Active and capable of digestion directly in GoTaq™ Green Master Mix. Stul is sensitive to overlapping dcm methylation. (AGG/CCT TCC\GGA)

  • Active and capable of digestion directly in GoTaq(R) Green Master Mix

‡Restriction enzyme buffer included indicated in parentheses; Cloning = Blue/White Cloning Qualified; Genome = Genome Qualified.

Specifications

Concentration 10 U/μL
Components 1 x 1mL 10X Buffer B, 1 x 150μL Bovine Serum Albumin, Acetylated, 1 x 400U StuI, 1 x 0.25mL Muti-Core™10X Buffer
Incubator Temperature 37°C
pH 7.4
For Use With (Application) Active and capable of digestion directly in GoTaq™ Green Master Mix
Storage Buffer 10mM Tris HCl (pH 7.4), 50mM NaCl, 0.1mM EDTA, 1mM DTT, 0.5mg/mL BSA, 50% Glycerol
Content And Storage -30°C to -10°C
Quantity 400 U
Cut Site AGG.CCT TCC.GGA
Product Type StuI
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.