Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Promega RET Kinase Enzyme System

Catalog No. PRV3761 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Quantity:
1/Ea.
10 μg
Product Type:
ADP-Glo Kinase Assay and RET Kinase System
RET Kinase Enzyme System
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Product Type
PRV3761 10 μg RET Kinase Enzyme System
PRV8061 1/Ea. ADP-Glo Kinase Assay and RET Kinase System
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. PRV3761 Supplier Promega Supplier No. V3761
Add to Cart
Add to Cart

Includes

RET Kinase Assay:

  • Kinase: Recombinant Human RET, 10μg (amino acids 658 to end), MW: ≅74kDa
  • Substrate: IGF1Rtide (KKKSPGEYVNIEFG) derived from human IRS-1 protein residues 891 to 902
  • Other: Reaction Buffer, DTT
ADP-Glo Kinase Assay: Reagent, buffer, substrate, Ultra Pure ATP (10mM), ADP (10mM)

Recombinant human RET (amino acids 658-end) was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag. RET gene codes for a transmembrane tyrosine kinase.

  • Easily Screen and Profile RET Kinase Inhibitors
  • Includes kinase, substrate and reaction buffer
  • Use with ADP-Glo™ Assay for bioluminescent detection of kinase activity

Specifications

Product Type RET Kinase Enzyme System
Molecular Weight (g/mol) 74
Content And Storage <-65°C
For Use With (Application) For both primary screening as well as Kinase selectivity profiling
Quantity 10 μg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.