Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Promega RET Kinase Enzyme System

Catalog No. p-4672305
Click to view available options
Quantity:
1/Ea.
10 μg
Product Type:
ADP-Glo Kinase Assay and RET Kinase System
RET Kinase Enzyme System

Includes

RET Kinase Assay:

  • Kinase: Recombinant Human RET, 10μg (amino acids 658 to end), MW: ≅74kDa
  • Substrate: IGF1Rtide (KKKSPGEYVNIEFG) derived from human IRS-1 protein residues 891 to 902
  • Other: Reaction Buffer, DTT
ADP-Glo Kinase Assay: Reagent, buffer, substrate, Ultra Pure ATP (10mM), ADP (10mM)

Recombinant human RET (amino acids 658-end) was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag. RET gene codes for a transmembrane tyrosine kinase.

  • Easily Screen and Profile RET Kinase Inhibitors
  • Includes kinase, substrate and reaction buffer
  • Use with ADP-Glo™ Assay for bioluminescent detection of kinase activity

Specifications

Product Type RET Kinase Enzyme System
Molecular Weight (g/mol) 74
Content And Storage <-65°C
For Use With (Application) For both primary screening as well as Kinase selectivity profiling
Quantity 10 μg
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.