Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Promega Reverse Transcription System 10X Buffer

Catalog No. PRA3561 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Concentration:
10X
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Concentration
PR-A3561 10X
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PR-A3561 Supplier Promega Supplier No. A3561
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Includes

Necessary enzymes, primers, and buffers; dNTPs; Nuclease-free water

The Reverse Transcription System provides reagents to efficiently reverse transcribe RNA into cDNA in fifteen minutes.

  • Convenient System for Reverse Transcription Using AMV Reverse Transcriptase

U.S. Patent 5,552,302; Australian Patent 646803 and other patents.

Specifications

Concentration 10X
Content And Storage 1 x 1.4mL Reverse transcription 10X Buffer. Store at −30 to −10°C
Quantity 1.4mL
For Use With (Application) RT-PCR
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.