Learn More
Promega Reverse Transcription System
Catalog No. PRA3500 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
For Use With (Application):
RT-PCR
Catalog No. For Use With (Application)
PR-A3500 RT-PCR
1 options
Catalog No. PR-A3500 Supplier Promega Supplier No. A3500
Includes
Necessary enzymes, primers, and buffers; dNTPs; Nuclease-free water
The Reverse Transcription System provides reagents to efficiently reverse transcribe RNA into cDNA in fifteen minutes.
  • Convenient System for Reverse Transcription Using AMV Reverse Transcriptase
U.S. Patent 5,552,302; Australian Patent 646803 and other patents.
Specifications
Content And Storage 1 x 1.2mL MgCl2, 1 x 1.4mL reverse transcription 10X Buffer, 1 x 50μg oligo(dT)15 primer, 1 x 320μL dNTP mix, 1 x 50μg random primers, 1 x 5μg 1.2kb kanamycin positive control RNA, 1 x 1500u AMV Reverse Transcriptase (HC), 1 x 13mL Nuclease free Water. Store at −30 to −10°C.
Quantity 1 Kit, 100 rxns
For Use With (Application) RT-PCR