Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Promega rNTPs

Catalog No. PRE6031 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Product Type:
rATP
rCTP
rGTP
rNTP Set (rCTP, rATP, rUTP, rGTP)
rUTP
Quantity:
400 μL
4 x 400 μL
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type Quantity
PR-E6031 rGTP 400 μL
PR-E6000 rNTP Set (rCTP, rATP, rUTP, rGTP) 4 x 400 μL
PR-E6011 rATP 400 μL
PR-E6041 rCTP 400 μL
PR-E6021 rUTP 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. PR-E6031 Supplier Promega Supplier No. E6031
Add to Cart
Add to Cart

High-quality rNTPs. Provided at concentrations of 10mM or 100mM in water at neutral pH.

  • High-Quality Ribonucleoside Triphosphates (rNTPs)
  • 400μL rNTP supplied at 10mM or 100mM in water at neutral pH
  • Qualified for use with the Riboprobe(R) and HeLaScribe(R) Systems
  • Purity verified by HPLC

Specifications

Product Type rGTP
Content And Storage -30°C to -10°C
Concentration 100 mM
Quantity 400 μL
For Use With (Application) Provides a cell-free system for in-vitro transcription of eukaryotic genes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.