Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Promega Sequencing Grade Chymotrypsin

Catalog No. PRV1061 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Quantity:
100 μg
25 μg
Concentration:
0.5 to 1 μg/μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Concentration
PRV1061 25 μg 0.5 to 1 μg/μL
PRV1062 100 μg 0.5 to 1 μg/μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. PRV1061 Supplier Promega Supplier No. V1061
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Chymotrypsin is a highly-purified serine endopeptidase derived from bovine pancreas that preferentially hydrolyzes at the carboxyl side of aromatic amino acids: Tyr, Phe and Trp. Cleavage may also be observed, but at a lower rate, at Leu and Met.

  • Serine Protease that Cleaves at C-Terminus of Tyr, Phe and Trp
  • Optimal activity at pH 7.0-9.0.
  • Use alone or with other proteases for peptide mapping, peptide mass fingerprinting, MS/MS spectral matching
  • Use for digestion reactions in-solution or in-gel

Specifications

Concentration 0.5 to 1 μg/μL
Color Colorless
pH 7 to 9
For Use With (Application) Peptide mapping, Protein identification
Content And Storage 2°C to 10°C
Source Bovine pancreas
Quantity 25 μg
Product Type Chymotrypsin
Molecular Weight (g/mol) 25
Form Lyophilized Powder
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.