Learn More
Description
- Simple Measurement of Deacetylating Activities: Use a single-reagent-addition, homogeneous, add-mix-measure protocol for easy implementation from benchtop to screening.
- Highly Sensitive: Achieve 10- to 100-fold higher sensitivity than comparable fluorescence methods.
- Fast Data Acquisition: Measure maximum signal in as little as 10-15 minutes with persistent glow-type steady-state signal.
Specifications
Specifications
| Quantity | 35 μL |
| Content And Storage | -20°C |
| For Use With (Application) | Determining SIRT inhibitor or activator potency, SIRT Enzyme profiling |
| Kit Contents | 1 x 35μL, ASIRT-Glo™ control substrate |
| pH | 8 |
| Form | Fluid |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.