Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Promega SoftLink™ Soft Release Avidin Resin
Click to view available options
Quantity:
1 mL
5 mL
Description
- Reversible Binding of Biotinylated Proteins Including Antibodies
- Specific capture of biotinylated proteins with reversible binding under mild elution conditions
- Binding ability retained under a wide range of pHs, and low or high ionic strength solutions
- Regenerates at least 10 times without loss of binding capacity
- For research use only. Not for diagnostic or therapeutic use.
- Exclusively licensed under U.S. Patents 5,395,856; and 6,072,039 and other patents.
Specifications
Specifications
Flow Rate | 300 cm/hr. |
pH Range | 2 to 13 |
Support Type | Methacrylate Polymeric Gel filtration matrix |
Content And Storage | 2°C to 10°C |
Quantity | 1 mL |
For Use With (Application) | Isolation and purification of Biotinylated Antibodies and other Proteins |
Type | Resin |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction