Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Promega SP6 RNA Polymerase

Catalog No. p-3509028 Shop All Promega Corporation Products
Change view
Click to view available options
Quantity:
1000 U
5000 U
Concentration:
10 to 20 U/μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Concentration
PR-P1085 1000 U 10 to 20 U/μL
PRP1081 5000 U 10 to 20 U/μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. PR-P1085 Supplier Promega Supplier No. P1085
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

DNA-dependent RNA polymerase for in vitro transcription.

  • DNA-Dependent RNA Polymerase Specific for SP6 Promoter Sequences
  • Exhibits extremely high affinity and specificity for SP6 promoter sequences
  • >90% pure as determined by SDS polyacrylamide gel electrophoresis
  • Free of detectable levels of contaminating RNase and DNase activity (

Specifications

Concentration 10 to 20 U/μL
Color Colorless
Components 100mM DTT 5X Reaction Buffer, transcription optimized 5X Buffer: 200mM Tris HCl (pH 7.9 at 25°C), 30mM MgCl2, 10mM Spermidine, 50mM NaCl
Purity >90%
pH 7.5
For Use With (Application) Generation of templates for in-vitro translation, generation of probes for Nucleic Acid hybridization, generation of RNA processing Substrates, generation of Antisense RNA
Storage Buffer 20mM Potassium Phosphate (pH 7.7 at 25°C), 1mM EDTA, 10mM DTT, 100mM NaCl, 0.1% Triton™ X-100, 50% Glycerol
Content And Storage -30°C to -10°C
Source Recombinant E. coli strain
Quantity 1000 U
Product Type SP6 RNA Polymerase
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.