Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Promega StemElite™ Differentiation-Associated Transcripts
Specifications
Product Type | Primer Pair |
---|---|
Concentration | 20 X |
For Use With (Application) | Differentiation of Stem Cells, Real-time Detection and Quantification of RNA Expression Levels |
Specifications
Primer Pair | |
20 X | |
Differentiation of Stem Cells, Real-time Detection and Quantification of RNA Expression Levels |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title