Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Promega Streptavidin Alkaline Phosphatase

Catalog No. PRV5591 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Quantity:
0.5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PR-V5591 0.5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PR-V5591 Supplier Promega Supplier No. V5591
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Used for the detection of biotinylated molecules.

  • Quality tested to ensure optimal performance for detecting biotinylated molecules

Specifications

Color Colorless
pH 7
For Use With (Application) Detection of Biotinylated molecules
Content And Storage 2°C to 10°C
Quantity 0.5 mL
Product Type Streptavidin Alkaline Phosphatase
Form Liquid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.