Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Promega SYK Kinase Enzyme System

Catalog No. PRV3801 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Quantity:
1/Ea.
10 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PRV3801 10 μg
PRV8271 1/Ea.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. PRV3801 Supplier Promega Supplier No. V3801
Add to Cart
Add to Cart

Includes

  • Kinase: Recombinant Human SYK, 10μg (full length), MW: ∽100kDa
  • Substrate: Poly (4:1 Glu, Tyr) Peptide
  • Other: Reaction Buffer, DTT

Recombinant full-length human SYK was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag. SYK is a non-receptor protein tyrosine kinase that is widely expressed in hematopoietic cells.

  • Easily Screen and Profile SYK Kinase Inhibitors
  • Includes kinase, substrate and reaction buffer
  • Use with ADP-Glo™ Assay for bioluminescent detection of kinase activity

Specifications

Molecular Weight (g/mol) 100
Content And Storage <-65°C
Quantity 10 μg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.