Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Promega T4 Polynucleotide Kinase Buffer Pack

Catalog No. PRC1313 Shop All Promega Corporation Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
3 x 500 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PR-C1313 3 x 500 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PR-C1313 Supplier Promega Supplier No. C1313
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

T4 PNK catalyzes the transfer of the gamma-phosphate from ATP to the 5'-terminus of polynucleotides or to mononucleotides bearing a 5'-hydroxyl group.

  • Transfers the γ-Phosphate from ATP to the 5′ End of Nucleotides
  • Can be inactivated by heating at 68°C for 10 minutes
  • Blue/white cloning qualified, providing a higher level of quality control for enzymes used in cloning applications
  • Labels the 5′ end of ssDNA, dsDNA and RNA molecules for use as probes or for sequencing or DNA-protein footprinting
  • Supplied at a concentration of 5-10u/μL

Specifications

Color Colorless
Components 3 Tubes of T4 DNA Kinase 10X Reaction Buffer: 700mM Tris HCl (pH 7.6 at 25degC), 100mM MgCl2, 50mM DTT
pH 8
For Use With (Application) For T4 DNA Kinase Enzyme
Content And Storage -30°C to -10°C
Quantity 3 x 500 μL
Product Type 4 DNA Kinase Buffer Pack
Form Liquid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.