Learn More
Specifications
Specifications
| Concentration | 10X |
| Formulation | 2-Amino-2-(Hydroxymethyl)-1,3-Propanediol 1 to 5% |
| For Use With (Application) | ELISA |
| Quantity | 20mL |
| Storage Requirements | 4°C |
| Product Type | TGFbeta Sample 10x Buffer |
| For Use With (Equipment) | TGFbeta1 and TGFbeta2 Emax Immunoassay System |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.